missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat F(ab) anti-Mouse IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-51890
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot, SDS-Page
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:100-1:2000, ELISA, Immunohistochemistry 1:1000 - 1:5000, Dot Blot, SDS-Page | |
| Goat | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot, SDS-Page | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Goat F(ab) anti-Mouse IgG (H+L) Secondary Antibody was produced by repeated immunization with mouse IgG whole molecule in goat. | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction