missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Antibody (GT175) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.60 - € 351.75
Specifications
| Antigen | Goat IgG |
|---|---|
| Clone | GT175 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601808
|
Novus Biologicals
NBP2-50413-0.025ml |
0.025 mL |
€ 214.00 € 201.60 / 0.03mL Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603579
|
Novus Biologicals
NBP2-50413 |
0.1 mL |
€ 372.00 € 351.75 / 0.10mL Save € 20.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Goat IgG Monoclonal antibody specifically detects Goat IgG in Goat samples. It is validated for ImmunohistochemistrySpecifications
| Goat IgG | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Goat | |
| Immunoglobulin G | |
| IgG1 | |
| Protein A purified |
| GT175 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Goat immunoglobulins | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title