missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Antibody (GT175), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50413DL594
This item is not returnable.
View return policy
Description
Goat IgG Monoclonal antibody specifically detects Goat IgG in Goat samples. It is validated for Immunohistochemistry
Specifications
| Goat IgG | |
| Monoclonal | |
| DyLight 594 | |
| Immunoglobulin G | |
| Goat immunoglobulins | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| GT175 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Goat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction