missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat Serum Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44080
This item is not returnable.
View return policy
Description
Specifically detects Goat Serum in Goat samples, and it is validated for ELISA, Immunoprecipitation, Western Blotting
Goat Serum Polyclonal specifically detects Goat Serum in Rat, Goat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Goat Serum | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Goat Sera | |
| Goat Serum Antibody was produced by repeated immunizations with Goat serum proteins. (Uniprot: P85295) | |
| Delipidation and Defibrination | |
| RUO | |
| Reconstitute with 2.0 ml deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500 | |
| P85295 | |
| Rabbit | |
| 10.06 kDa | |
| 2 mL | |
| Primary | |
| Rat, Goat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction