missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GODZ Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06315APC
This item is not returnable.
View return policy
Description
GODZ Polyclonal antibody specifically detects GODZ in Human samples. It is validated for Western Blot
Specifications
| GODZ | |
| Polyclonal | |
| Western Blot | |
| DHHC domain containing 3, DKFZp313D2314, EC 2.3.1, EC 2.3.1.-, FLJ20209, FLJ45940, Protein DHHC1, ZDHC3, zinc finger, DHHC-type containing 3 | |
| Produced in rabbits immunized with E. coli-derived Human GODZ fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 51304 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction