Learn More
Gold for AAS 1g/L HCl matrix, Fisher Chemical™
Catalog Number 12923125
Click to view available options
Quantity:
100 mL
500 mL
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.97
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
CHEBI:30050
gold
[Au]
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Concentration 1g/L ±0.002g/L
Packaging Glass Bottle
Quantity 500 mL
Type Flame Single-element Standard Solution
Solvent or Matrix HCl
Physical Form Liquid
Chemical Name or Material Gold