Learn More
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Product Code. 10219174 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.2 mm
For Use With (Equipment):
Dual Column Installation
Single Column Installation
Single Column Installation (Cross Version)
Quantity:
10 Pack
2 Pack
Product Type:
Gold Inlet Base Seal
Siltek Inlet Base Seal
Unit Size:
Pack of 10
Pack of 2
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner For Use With (Equipment) Quantity Product Type unitSize
10219174 0.8 mm Single Column Installation (Cross Version) 2 Pack Gold Inlet Base Seal Pack of 2
10281665 0.8 mm Single Column Installation 10 Pack Gold Inlet Base Seal Pack of 10
10548605 0.8 mm Single Column Installation 2 Pack Gold Inlet Base Seal Pack of 2
10558605 1.2 mm Dual Column Installation 10 Pack Gold Inlet Base Seal Pack of 10
10703996 1.2 mm Dual Column Installation 2 Pack Gold Inlet Base Seal Pack of 2
10371335 0.8 mm Single Column Installation (Cross Version) 10 Pack Gold Inlet Base Seal Pack of 10
10534655 0.8 mm Single Column Installation 10 Pack Siltek Inlet Base Seal Pack of 10
10781947 0.8 mm Single Column Installation 2 Pack Siltek Inlet Base Seal Pack of 2
10575425 0.8 mm Single Column Installation (Cross Version) 2 Pack Siltek Inlet Base Seal Pack of 2
10417714 0.8 mm Single Column Installation (Cross Version) 10 Pack Siltek Inlet Base Seal Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10219174 Supplier Thermo Scientific™ Supplier No. 290GA083
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 Pack
Product Type Gold Inlet Base Seal
Diameter (Metric) Inner 0.8 mm
For Use With (Equipment) Single Column Installation (Cross Version)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.