missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
€ 425.00 - € 557.00
Specifications
| Antigen | GOLGA5 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436970
|
Novus Biologicals
NBP1-83352-25ul |
25 μL |
€ 425.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18231548
|
Novus Biologicals
NBP1-83352 |
0.1 mL |
€ 557.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOLGA5 Polyclonal specifically detects GOLGA5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GOLGA5 | |
| Polyclonal | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9950 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SSVNPSVTTIKTIEENSFGSQTHEAASNSDSSHEGQEESSKENVSSNAACPDHTPTPNDDGKSHELSNLRLENQLLRNEVQSLNQEMASLLQRSKETQEELNKARARVEKWNADHSKSDRMTRGLRAQVDDLTEAVAAKDSQLAVLKV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Cell proliferation-inducing gene 31 protein, golgi autoantigen, golgin subfamily a, 5, golgin A5, Golgin subfamily A member 5, Golgin-84, GOLIM5, Protein Ret-II, PTC5, RET-fused gene 5 protein, ret-II, RETII, rfg5, RFG5golgi integral membrane protein 5 | |
| GOLGA5 | |
| IgG | |
| Affinity Purified | |
| Specificity of human GOLGA5 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title