missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGB1/Giantin Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22323APC
This item is not returnable.
View return policy
Description
GOLGB1/Giantin Polyclonal antibody specifically detects GOLGB1/Giantin in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| GOLGB1/Giantin | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| 372 kDa Golgi complex-associated protein, GCP, GCP372GIANTIN, giantin, golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal)1,macrogolgin, golgi integral membrane protein 1, golgin B1, golgin B1, golgi integral membrane protein, Golgin subfamily B member 1, GOLIM1, Macrogolgin | |
| The immunogen this antibody was made to, maps to a region between residue 1975 to 2025 of human Golgin B1 using the numbering given in entry NP_004478.3 (GeneID 2804). | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 2804 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction