missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (AE-6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11427AF350
This item is not returnable.
View return policy
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Golgi Complex | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| AE-6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
| SU-DHL-1 large cell lymphoma cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction