missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgi protein 58k Recombinant Rabbit Monoclonal Antibody (3X2O4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555273
This item is not returnable.
View return policy
Description
Golgi protein 58k Recombinant Monoclonal Antibody for Western Blot, ELISA
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
| Golgi protein 58k | |
| Recombinant Monoclonal | |
| 0.26 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O88618, O95954, Q91XD4 | |
| Ftcd | |
| Recombinant protein of human FTCD. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3X2O4 | |
| Unconjugated | |
| Ftcd | |
| 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10841, 14317, 89833 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction