missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgin-97 Recombinant Rabbit Monoclonal Antibody (23GB1800), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553128
This item is not returnable.
View return policy
Description
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
Specifications
| Golgin-97 | |
| Recombinant Monoclonal | |
| 1.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92805, Q9CW79 | |
| GOLGA1 | |
| A synthesized peptide derived from human Golgin 97 (1-57AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1800 | |
| Unconjugated | |
| GOLGA1 | |
| 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2800, 311919, 76899 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction