missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (015), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90164MFV500
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Flow Cytometry
Specifications
| GOLM1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 015 | |
| Flow Cytometry | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human GOLM1 (Q8NBJ4-1; Met1-Leu401). | |
| 0.1 mL | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction