missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (12), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12533UV
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| GOLM1 | |
| Monoclonal | |
| DyLight 350 | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Recombinant Human GOLM1 Protein | |
| 100 μg | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction