missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (OTI4B12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71916CL1
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human,Monkey,Primate samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| GOLM1 | |
| Monoclonal | |
| CoraFluor 1 | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Full length human recombinant protein of human GOLM1(NP_808800) produced in HEK293T cell. | |
| 0.1 mL | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| OTI4B12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey, Primate | |
| Purified |
Product Content Correction
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto