missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Antibody (OTI4B12), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71916JF549
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal specifically detects GOLM1 in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GOLM1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GOLM1 | |
| Full length recombinant protein of human GOLM1 (NP_808800) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Primate, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4B12 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| Mouse | |
| Affinity Purified | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction