missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLM1 Mouse anti-Human, Clone: OTI6C9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504113
This item is not returnable.
View return policy
Description
GOLPH2, also known as GOLM1, is a Golgi phosphoprotein that has a short cytoplasmic N-terminal domain, a membrane spanning region, and a longer C-terminal domain. It was initially identified as a possible marker for Alzheimer′s disease, although later studies have demonstrated that the GOLPH2 gene does not contribute to risk of this disease. GOLPH2 expression has been reported higher in prostate cancer tissues compared to normal prostate tissue, suggesting that GOLPH2 can be used as an additional positive marker for tissue-based diagnosis of prostate cancer. It has been suggested that GOLPH2 expression in hepatocellular carcinomas (HCCs) and serum may also serve as tumor markers for HCCs.Specifications
| GOLM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NBJ4 | |
| GOLM1 | |
| Full length human recombit protein of human GOLM1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C9 | |
| Unconjugated | |
| GOLM1 | |
| C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51280 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction