missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18486AF647
This item is not returnable.
View return policy
Description
GOLPH3 Polyclonal antibody specifically detects GOLPH3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GOLPH3 | |
| Polyclonal | |
| Western Blot | |
| Coat protein GPP34, FLJ90675, Golgi phosphoprotein 3, golgi phosphoprotein 3 (coat-protein), golgi protein, golgi-associated protein, GOPP1, GPP34coat-protein, MIDASgolgi peripheral membrane protein 1, 34 kDa, Mitochondrial DNA absence factor | |
| Recognizes region between residues Between between 1 and 50.. | |
| 0.1 mL | |
| Cancer | |
| 64083 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction