missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH3 Polyclonal Antibody

GOLPH3 Polyclonal Antibody
Brand: Invitrogen PA598901
This item is not returnable.
View return policy
Description
GOLPH3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Specifications
| GOLPH3 | |
| Polyclonal | |
| Unconjugated | |
| Golph3 | |
| 4733401N08Rik; 5730410D03Rik; AW413496; coat protein GPP34; coat-protein; Gmx33; golgi peripheral membrane protein 1, 34 kDa; Golgi phosphoprotein 3; golgi phosphoprotein 3 (coat-protein); golgi protein; golgi-associated protein; GOLPH3; GOPP1; GPP34; MIDAS; Mitochondrial DNA absence factor; trans-Golgi protein GMx33; Vps74 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 64083 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Western Blot | |
| 3.99 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H4A6 | |
| Golph3 | |
| Recombinant Human Golgi phosphoprotein 3 protein (16-150AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction