missing translation for 'onlineSavingsMsg'
Learn More

Golvatinib, MedChemExpress

Product Code. 16498237
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498237 5 mg 5mg
16486096 100 mg 100mg
16421707 50 mg 50mg
16415538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498237 Supplier Medchem Express Supplier No. HY13068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Golvatinib (E-7050) is a potent dual inhibitor of both c-Met and VEGFR2 kinases with IC50s of 14 and 16 nM, respectively.

Golvatinib (E-7050) is a potent dual inhibitor of both c-Met and VEGFR2 kinases with IC50s of 14 and 16 nM, respectively.

Specifications

Chemical Name or Material Golvatinib
CAS 928037-13-2
Color White
Purity Grade Notes Research
Molecular Formula C33H37F2N7O4
Quantity 5 mg
Synonym E-7050
Solubility Information DMSO : 50 mg/mL (78.90 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C=C1F)OC2=CC(NC(N3CCC(CC3)N4CCN(CC4)C)=O)=NC=C2)C5(CC5)C(NC6=CC=C(C=C6)F)=O
Molecular Weight (g/mol) 633.69
Formula Weight 633.69
Percent Purity 99.15%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.