missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (OTI1D7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72409AF594
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Goosecoid | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GSC | |
| Recombinant protein fragment corresponding to amino acids 107-257 of human Goosecoid (NP_776248) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1D7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| goosecoid homeobox, homeobox protein goosecoid | |
| Mouse | |
| 0.1 mL | |
| 145258 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction