missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (OTI1D7), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72409PECY55
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Goosecoid | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| GSC | |
| Recombinant protein fragment corresponding to amino acids 107-257 of human Goosecoid (NP_776248) produced in E.coli. | |
| 0.1 mL | |
| 145258.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1D7 | |
| Flow Cytometry | |
| goosecoid homeobox, homeobox protein goosecoid | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction