Learn More
Gosselin™ Square HDPE 150mL Bottles
Product Code. 11703034
Click to view available options
Closure Type:
Double Screw Cap
Screw Cap
Screw Cap with Wad
Single Screw Cap
Closure Material:
HDPE
LDPE
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 300
Product Code. 11703034
Brand: Gosselin™ LC150A07
Only null left
Add to basket
Add to basket
Provides excellent storage for liquids and solids in the laboratory. Gosselin™ Square HDPE 150mL Bottles come with either a screw cap with inner lip, with separate seal or with wad.
  • Large mouth
  • Break resistant
  • Sterile or aseptic by irradiation
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 150 mL
Color Natural
Description Square
Diameter (Metric) Inner Neck 37 mm
Material HDPE
Length (Metric) 50mm
Sterility Non-sterile
Width (Metric) 50mm
Closure Color Red
Certifications/Compliance EC N°1935/2004
Closure Included Yes
Disposable Single-use
Mouth Wide
Height (Metric) 95 mm
Closure Liner Tresylene
Closure Material HDPE
Closure Type Screw Cap with Wad