Learn More
Gosselin™ Square HDPE Bottles
Click to view available options
Closure Color:
Red
White
Diameter (Metric) Inner Neck:
22 mm
28 mm
29 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 77
Description
- Break resistant
- With molded graduations
- With either a screw cap with shaped seal or hinged cap with inner lip and safety ring
- Sterilized by irradiation with peel-away sterilization certificate on the outer carton
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 22 mm |
| Includes | White LDPE screw cap with shaped seal |
| Material | HDPE |
| Length (Metric) | 70mm |
| Quantity | 100/Carton |
| Sterility | Non-sterile |
| Width (Metric) | 70mm |
| Closure Color | White |
| Show More |