missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOT2 Antibody (8B11), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59428IR
This item is not returnable.
View return policy
Description
GOT2 Monoclonal antibody specifically detects GOT2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| GOT2 | |
| Monoclonal | |
| DyLight 755 | |
| aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
| Recombinant human GOT2 (30-430aa) purified from E.coli. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 2806 | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 8B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction