missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp130/CD130 Antibody (28126), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB228D
This item is not returnable.
View return policy
Description
gp130/CD130 Monoclonal antibody specifically detects gp130/CD130 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF
Specifications
| gp130/CD130 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, CyTOF | |
| 28126 | |
| Flow Cytometry, Neutralization, CyTOF-ready | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 extracellular domain | |
| 0.1 mL | |
| Cancer, Immunology | |
| 3572 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction