missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp130/CD130 Antibody (MM0318-9X23), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12344PECY7
This item is not returnable.
View return policy
Description
gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Flow Cytometry.
Specifications
| gp130/CD130 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| IL6ST | |
| Human recombinant gp130/CD130 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| MM0318-9X23 | |
| Flow Cytometry | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| Mouse | |
| Protein G purified | |
| Cancer, Immunology | |
| 3572.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction