missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP1BB Antibody (Gi27), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-89008
This item is not returnable.
View return policy
Description
GP1BB Monoclonal specifically detects GP1BB in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GP1BB | |
| Monoclonal | |
| Unconjugated | |
| 100ug of purified antibody in 200 ul PBS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunoprecipitation | |
| Gi27 | |
| Western Blot 1:20, Immunoprecipitation | |
| antigen CD42b-beta, BDPLT1, BS, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta | |
| Human platelets. | |
| RUO | |
| 2812 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction