missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP1BB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33169
This item is not returnable.
View return policy
Description
GP1BB Polyclonal specifically detects GP1BB in Human, Mouse samples. It is validated for Western Blot.
Specifications
| GP1BB | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GP1BB | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human CD42c. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2812 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 0.75mg/mL | |
| Western Blot 1:500-1:3000 | |
| antigen CD42b-beta, BDPLT1, BS, glycoprotein Ib (platelet), beta polypeptide, glycoprotein Ib platelet beta subunit, GPIBB, GPIbbeta, GPIb-beta, nuclear localization signal deleted in velocardiofacial syndrome, platelet glycoprotein Ib beta chain, platelet membrane glycoprotein Ib beta, truncated platelet membrane glycoprotein Ib beta | |
| Rabbit | |
| 22 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction