missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GP2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA592932

Product Code. 16508311

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CSSILQTEER NWVSVTSPVQ ASACRNILER NQTHAIYKNT LSLVNDFIIR DTILNINFQC AYPLDMKVSL QAALQPIVSS LNVSVDGNGE FIVRMALFQD QNYTNPYEGD AVELSVESVL YVGAILEQGD TSRFNLVLRN CYATPTEDKA DLVKYFIIRN SCSNQRDSTI HVEENGQSSE SRFSVQMFMF AGHYDLVFLH CEIHLCDSLN EQCQPSCSRS QVRSEVPAID LARVLDLGPI TRRGAQSPGV MNGTPSTAGF LVAWPMVLLT VLLAWLF; Positive Samples: U-251MG; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.

GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GP2
Polyclonal
Unconjugated
GP2
2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2
Rabbit
Affinity Chromatography
RUO
2813
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55259
GP2
Recombinant fusion protein containing a sequence corresponding to amino acids 111-387 of human GP2 (NP_0010072432).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.