missing translation for 'onlineSavingsMsg'
Learn More

GP73/GOLPH2 Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

€ 149.00 - € 439.00

Specifications

Antigen GP73/GOLPH2
Clone 1I12
Concentration 500 μg/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236836
View Documents
Proteintech
81893-1-RR-20UL
20 μL
€ 149.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234783
View Documents
Proteintech
81893-1-RR-100UL
100 μL
€ 439.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GP73/GOLPH2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human GP73/GOLPH2. This antibody recognizes human antigen. The GP73/GOLPH2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.
Specifications

Specifications

GP73/GOLPH2
500 μg/mL
Recombinant Monoclonal
Liquid
RUO
51280
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1I12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Rabbit
Human
C9orf155; FLJ22634; FLJ23608; GOLPH2; GP73; PSEC0257; bA379P1.3
IgG
Protein A purification
Recombinant Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.