missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp96/HSP90B1/GRP94 Antibody (CL2647), Novus Biologicals™
Mouse Monoclonal Antibody
€ 289.00 - € 518.00
Specifications
| Antigen | gp96/HSP90B1/GRP94 |
|---|---|
| Clone | CL2647 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613969
|
Novus Biologicals
NBP2-42379-25ul |
25 μL |
€ 289.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685876
|
Novus Biologicals
NBP2-42379 |
0.1 mL |
€ 518.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| gp96/HSP90B1/GRP94 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 94 kDa glucose-regulated protein, endothelial cell (HBMEC) glycoprotein, glucose regulated protein, 94 kDa, GP96ECGP, GRP-94, GRP94gp96 homolog, Heat shock protein 90 kDa beta member 1, heat shock protein 90kDa beta (Grp94), member 1, TRA1endoplasmin, tumor rejection antigen (gp96) 1, Tumor rejection antigen 1, Tumor rejection antigen-1 (gp96) | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SGNMERIMKAQAYQTGKDISTNYYASQKKTFEINPRHPLIRDMLRRIKEDEDDKTVLDLAVVLFETATLRSGYLLPDTKAYGDRIERMLRLSLNIDPDAKVEEEPEEEPEETAEDTTEDTEQDEDEEMDVGTDEEEETAKESTA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL2647 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| PBS (pH 7.2), 40% Glycerol | |
| 7184 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title