missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPATCH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560434
This item is not returnable.
View return policy
Description
Immunogen sequence: TLKASNFKPF AKDPEKQKRY DEFLVHMKQG QKDALERCLD PSMTEWERGR ERDEFARAAL LYASSHSTLS SRFTHAKEED DSDQVEVPRD QENDVGDKQS AVKMKMFGKL TR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifications
| GPATCH1 | |
| Polyclonal | |
| Unconjugated | |
| GPATCH1 | |
| 1300003A17Rik; ECGP; evolutionarily conserved G patch domain containing; evolutionarily conserved G-patch domain containing; evolutionarily conserved G-patch domain-containing protein; G patch domain containing 1; G patch domain-containing protein 1; GPATC1; G-patch domain containing 1; Gpatch1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 292810, 55094, 67471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRR8, Q9DBM1 | |
| GPATCH1 | |
| Recombinant protein corresponding to Human GPATCH1. Recombinant protein control fragment (Product #RP-98881). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction