missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPC1 Monoclonal Antibody (OTI3G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812434S
This item is not returnable.
View return policy
Description
GPC1 Monoclonal antibody specifically detects GPC1 in Human samples. It is validated for Western BlotSpecifications
| GPC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35052 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2817 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3G9 | |
| Unconjugated | |
| GPC1 | |
| AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1 | |
| Human recombinant protein fragment corresponding to amino acids 24-530 of human GPC1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction