missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPC3, anti-Human, Clone: GPC3/1534R, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00280.100ug
This item is not returnable.
View return policy
Specifications
| GPC3 | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS. | |
| DGSX/OCI-5/SDYS/SGB/SGBS/SGBS1 | |
| Original antibody is raised against recombinant protein corresponding to full length human GPC3. | |
| RUO | |
| 2719 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GPC3/1534R | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| glypican 3 | |
| GPC3 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction