missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531051
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Mouse (91%), Rat (91%), Pig (91%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the NAD-dependent glycerol-3-phosphate dehydrogenase family. The encoded protein plays a critical role in carbohydrate and lipid metabolism by catalyzing the reversible conversion of dihydroxyacetone phosphate (DHAP) and reduced nicotine adenine dinucleotide (NADH) to glycerol-3-phosphate (G3P) and NAD+. The encoded cytosolic protein and mitochondrial glycerol-3-phosphate dehydrogenase also form a glycerol phosphate shuttle that facilitates the transfer of reducing equivalents from the cytosol to mitochondria. Mutations in this gene are a cause of transient infantile hypertriglyceridemia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| GPD1 | |
| Polyclonal | |
| Unconjugated | |
| Gpd1 | |
| AI747587; Gdc1; Gdc-1; glycerol 3-phosphate dehydrogenase; glycerol phosphate dehydrogenase 1, cytoplasmic adult; Glycerol-3-phosphate dehydrogenase [NAD(+)], cytoplasmic; glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic; glycerol-3-phosphate dehydrogenase 1; glycerol-3-phosphate dehydrogenase 1 (soluble); glycerolphosphate dehydrogenase 1, cytoplasmic adult; glycerophosphate dehydrogenase; Gpd1; Gpd3; GPD-C; GPDH; GPDH-C; HTGTI; Kiaa4010; mKIAA4010 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2819, 60666 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.51 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O35077, P21695 | |
| Gpd1 | |
| Recombinant protein encompassing a sequence within the center region of human GPD1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction