missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPD1L Antibody (14E2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59439AF532
This item is not returnable.
View return policy
Description
GPD1L Monoclonal antibody specifically detects GPD1L in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| GPD1L | |
| Monoclonal | |
| Alexa Fluor 532 | |
| EC 1.1.1, EC 1.1.1.8, glycerol-3-phosphate dehydrogenase 1-like, glycerol-3-phosphate dehydrogenase 1-like protein, KIAA0089GPD1-L | |
| Recombinant human GPD1L (1-351aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 23171 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1400 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction