missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100148
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPD2.
Mitochondrial glycerophosphate dehydrogenase (EC 1. 1. 99. 5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.
Specifications
| GPD2 | |
| Polyclonal | |
| Unconjugated | |
| Gpd2 | |
| AA408484; AI448216; AU021455; AW494132; FAD-linked glycerol-3-phosphate dehydrogenase; GDH2; Gdm1; glycerol phosphate dehydrogenase 1, mitochondrial; glycerol phosphate dehydrogenase 2, mitochondrial; Glycerol-3-phosophate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2; glycerol-3-phosphate dehydrogenase 2 (mitochondrial); glycerol-3-phosphate dehydrogenase 2, mitochondrial; glycerol-3-phosphate dehydrogenase, mitochondrial; Gpd2; GPDH; GPDH-M; GPDM; GPD-M; mGPDH; mitochondrial glycerol-3-phosphate dehydrogenase; mitochondrial glycerophosphate dehydrogenase; mtGPD; mtGPDH; mtGPDH gene, promoter region and alternative transcripts; Protein TISP38; testicular tissue protein Li 76; TISP38 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14571, 25062, 2820 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35571, P43304, Q64521 | |
| Gpd2 | |
| A synthesized peptide derived from human GPD2(Accession P43304), corresponding to amino acid residues S82-E132. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction