missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPER/GPR30 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS4272
This item is not returnable.
View return policy
Description
GPER/GPR30 Polyclonal antibody specifically detects GPER/GPR30 in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| GPER/GPR30 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| CEPRG-protein coupled receptor 30, Chemoattractant receptor-like 2, chemokine receptor-like 2, CMKRL2MGC99678, constitutively expressed peptide-like receptor, DRY12IL8-related receptor DRY12, FEG-1mER, Flow-induced endothelial G-protein coupled receptor 1, G protein-coupled estrogen receptor 1, G protein-coupled receptor 30, GPCR-BR, GPR30GPCR-Br, G-protein coupled estrogen receptor 1, heptahelix receptor, LERGU, LERGU2, leucine rich protein in GPR30 3'UTR, LyGPR, Lymphocyte-derived G-protein coupled receptor, Membrane estrogen receptor | |
| Synthetic 18 amino acid peptide from C-terminus of human GPER/GPR30. | |
| 0.05 mL | |
| Breast Cancer, Cancer, GPCR | |
| 2852 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 5-10 μg/mL, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Q99527 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction