Learn More
GPHA2 Antibody (06), Alexa Fluor™ 532 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06201AF532
Product Code. 18560002
  • € 423.00 / 0.10mL
Description
GPHA2 Monoclonal antibody specifically detects GPHA2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPHA2
Monoclonal
Alexa Fluor 532
A2, glycoprotein alpha 2, glycoprotein hormone alpha 2, glycoprotein hormone alpha-2, GPA2Thyrostimulin subunit alpha, MGC126572, Putative secreted protein Zsig51, ZSIG51cysteine knot protein
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPHA2 (Q96T91; Met1-Tyr129).
0.1 mL
Primary
Human
Purified
ELISA
06
50mM Sodium Borate
Mouse
Protein A purified
RUO
170589
Store at 4°C in the dark.