missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPI7 Polyclonal antibody specifically detects GPI7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GPI7 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | DKFZp434A1810, DKFZp434C1712, EC 2.-, FLJ20265, FLJ39925, GPI ethanolamine phosphate transferase 2, GPI7 homolog, GPI7DKFZp434M1131, hGPI7, LAS21, MGC131903, phosphatidylinositol glycan anchor biosynthesis, class G, phosphatidylinositol glycan, class G, Phosphatidylinositol-glycan biosynthesis class G protein, PIG-G, PRO4405, RLGS1930 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human GPI7 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?