missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-41539JF635
This item is not returnable.
View return policy
Description
GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| GPIHBP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 635 | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2] | |
| 0.1 mL | |
| Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 338328 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction