missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97041-100ul
This item is not returnable.
View return policy
Description
GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human samples. It is validated for ELISA
Specifications
| GPIHBP1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| Produced in rabbits immunized with purified, recombinant Human GPIHBP1 (Accession#: AAH63857.1; Met1-Ser160) | |
| 100 μg | |
| Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 338328 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction