missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPIHBP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11591832
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11591832 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11591832 Supplier Invitrogen™ Supplier No. PA116977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Suggested positive control: mouse kidney lysate or protein.

GPI-anchored HDL-binding protein 1 (Gpihbp1) is a novel protein that may be involved in the regulation of the delivery of fats to cells for energy and storage. Digested fats travel to the small intestine, where they are packaged into chylomicrons (particles filled with triglycerides). Chylomicrons then travel through the bloodstream and deliver triglycerides to tissues that are hungry for fuel or to adipose tissue for energy storage. Triglycerides are broken down or hydrolyzed by the enzyme lipoprotein lipase (LpL). The triglyceride breakdown products are then taken up and used by cells. Gpihbp1 is the molecule in capillaries that facilitates the capture of chylomicrons and facilitates the interaction with LpL. It has been shown that fats in the bloodstream are not readily metabolized in the absence of GPIHBP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPIHBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% sodium azide
Gene GPIHBP1
Gene Accession No. Q8IV16, Q9D1N2
Gene Alias 1110002J19Rik; endothelial cell LPL transporter; glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1; glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI anchored high density lipoprotein binding protein 1; GPI-anchored HDL-binding protein 1; Gpihbp1; GPI-HBP1; Hbp1; HDLBP; HDLBP1; High density lipoprotein-binding protein 1; HYPL1D; RGD1564237
Gene Symbols GPIHBP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338328, 68453
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.