missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIP137 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22237AF750
This item is not returnable.
View return policy
Description
GPIP137 Polyclonal antibody specifically detects GPIP137 in Human, Mouse samples. It is validated for Western Blot
Specifications
| GPIP137 | |
| Polyclonal | |
| Western Blot | |
| activation/proliferation-associated protein 1, caprin 1, caprin-1, cell cycle associated protein 1, Cell cycle-associated protein 1, Cytoplasmic activation- and proliferation-associated protein 1, cytoplasmic activation/proliferation-associated protein-1, GPI-anchored membrane protein 1GPIP137, GPI-p137, M11S1GPI-anchored protein p137, Membrane component chromosome 11 surface marker 1, membrane component, chromosome 11, surface marker 1, p137GPI, RNA granule protein 105, RNG105GPIAP1 | |
| The immunogen this antibody was made to, maps to a region between residue 450 and 500 of human Cell Cycle Associated Protein 1 using the numbering given in entry NP_005889.3 (GeneID 4076). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4076 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction