Learn More
Novus Biologicals™ GPIP137 Antibody (Toray patent anti-Caprin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28615-50ug
 View more versions of this product
Product Code. 30048811
  • € 213.00 / 50µg
Description
GPIP137 Monoclonal antibody specifically detects GPIP137 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPIP137
Monoclonal
Unconjugated
Q14444
Human
Protein A purified
RUO
4076
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Toray patent anti-Caprin-1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
activation/proliferation-associated protein 1, caprin 1, caprin-1, cell cycle associated protein 1, Cell cycle-associated protein 1, Cytoplasmic activation- and proliferation-associated protein 1, cytoplasmic activation/proliferation-associated protein-1, GPI-anchored membrane protein 1GPIP137, GPI-p137, M11S1GPI-anchored protein p137, Membrane component chromosome 11 surface marker 1, membrane component, chromosome 11, surface marker 1, p137GPI, RNA granule protein 105, RNG105GPIAP1
CAPRIN1
50 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.