missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ GPN 200 Series Natural Form A LDPE Cylindrical Protective Cap

Product Code. 10013994
Change view
Click to view available options
Length (Metric):
6 mm
16 mm
20 mm
25 mm
27 mm
30 mm
33 mm
35 mm
91 mm
Diameter (Metric) Inside:
5 mm
7 mm
8 mm
10 mm
12 mm
20 mm
27 mm
35 mm
42 mm
Unit Size:
Pack of 100
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Inside unitSize
10013994 16 mm 8 mm Pack of 100
10586244 33 mm 10 mm Pack of 100
10014044 6 mm 10 mm Pack of 100
10013774 35 mm 12 mm Pack of 100
10328724 27 mm 42 mm Pack of 100
10697785 20 mm 20 mm Pack of 100
10165294 20 mm 5 mm Pack of 100
10155214 25 mm 35 mm Pack of 100
10369004 20 mm 27 mm Pack of 100
10738565 91 mm 7 mm Pack of 100
10207214 30 mm 8 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10013994 Supplier Poeppelmann™ Supplier No. 20008160000
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Natural
Diameter (Metric) Inside 8 mm
For Use With (Application) For protection of threaded studs, pipe ends and shafts
Length (Metric) 16 mm
Quantity 100 Pack
Material LDPE
Type Protective Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.