Learn More
Poeppelmann™ KAPSTO™ GPN 260 Square Plugs
Featuring distinctive external ribs to provide a secure press fit. Poeppelmann™ KAPSTO™ GPN 260 Square Plugs are manufactured from black PE-LLD plastic.
Brand:  Poeppelmann™ 26020200000
Additional Details : Weight : 0.15300kg
 View more versions of this product
Product Code. 10073744
  • € 15.45 / Pack of 100
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • For closing of section tubes
  • Reinforced head area for higher demands
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications
Specifications
Wall thickness: 1.5 - 2 mm
Square Plugs
15mm
20mm
LDPE
20 mm
Black
For closing of section tubes
20 x 20 x 15mm
100/ck.
For use on profile tubing, with distinctive external ribs providing a secure press fit
Square