missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ GPN 260 Series Black LDPE Square Plugs
Specifications
Specifications
| Color | Black |
| For Use With (Application) | For closing of section tubes |
| Length (Metric) | 100 mm |
| Quantity | 25 Pack |
| Width (Metric) | 100 mm |
| Material | LDPE |
| Type | Square Plugs |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?