missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs

Featuring distinctive external ribs to provide a secure press fit. Poeppelmann™ KAPSTO™ GPN 270 Rectangular Plugs are manufactured from black PE-LLD plastic.

Brand:  Poeppelmann™ 27080440000

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10083714

  • € 59.10 / Pack of 25

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

  • For closing of section tubes
  • Reinforced head area for higher demands
  • High visual appeal due to grainded top surface finish and chamfered edges
Specifications

Specifications

Rectangular Closure Plug
80 x 40 mm
For closing of profile pipes
19 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers